This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PPIG SIRNA Assays ›
Gene Symbol:
PPIG
Gene Name:
peptidylprolyl isomerase G
Gene Aliases:
CARS-Cyp, CYP, SCAF10, SRCyp
Chromosome Location:
Chr.2: 169584340 - 169641406 on Build GRCh38
UniGene ID:
Hs.470544
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 107533

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPIG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004792.2 NP_004783.2 6, 7 505
XM_005246966.1 XP_005247023.1 6, 7 462
XM_005246967.1 XP_005247024.1 6, 7 509
XM_017005302.1 XP_016860791.1 4, 5 301
GenBank mRNA AK307552.1 - 3, 4 296
BC001555.1 - 7, 8 465
BC063711.1 - 6, 7 607
BC073875.1 - 5, 6 360
BC109373.1 - 6, 7 509
U40763.1 - 7, 8 770
X99717.1 - 6, 7 442

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
RNA splicing
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline