siRNA Details
| Gene Symbol: SLC30A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006345.3 | NP_006336.3 | 18 | 2640 | ||
| GenBank mRNA | AF006621.2 | - | 18 | 2614 | ||
| AK225470.1 | - | 18 | 2577 | |||
| AY319413.1 | - | 19 | 2519 | |||
| BC000240.1 | - | 18 | 2602 | |||
| BC007732.2 | - | 18 | 2601 | 4109702 | ||
| BC016949.1 | - | 18 | 2619 | 3850521 | ||
| BC022981.1 | - | 14 | 2031 | |||

SLC30A9
Genomic Map