This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FCER2 SIRNA Assays ›
Gene Symbol:
FCER2
Gene Name:
Fc fragment of IgE receptor II
Gene Aliases:
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Chromosome Location:
Chr.19: 7688757 - 7702755 on Build GRCh38
UniGene ID:
Hs.465778
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 10816

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FCER2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207019.2 NP_001193948.2 5 359
NM_001220500.1 NP_001207429.1 6 495
NM_002002.4 NP_001993.2 6 491
XM_005272462.4 XP_005272519.1 6 1096
GenBank mRNA AK310071.1 - 6 480
AK310707.1 - 6 460
BC014108.2 - 6 456 4309266
BC062591.1 - 6 449 5216918
BC064417.1 - 5 773
M14766.1 - 6 463
M15059.1 - 6 491
X04772.1 - 6 491

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of humoral immune response mediated by circulating immunoglobulin
Notch signaling pathway
positive regulation of nitric-oxide synthase activity
positive regulation of killing of cells of other organism
positive regulation of nitric-oxide synthase biosynthetic process
integrin binding
protein binding
IgE binding
carbohydrate binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline