This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TARDBP SIRNA Assays ›
Gene Symbol:
TARDBP
Gene Name:
TAR DNA binding protein
Gene Aliases:
ALS10, TDP-43
Chromosome Location:
Chr.1: 11012622 - 11030528 on Build GRCh38
UniGene ID:
Hs.300624
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 108256

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TARDBP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007375.3 NP_031401.1 6 1000
XM_017000863.1 XP_016856352.1 6 988
XM_017000864.1 XP_016856353.1 6 988
XM_017000865.1 XP_016856354.1 6 988
XM_017000866.1 XP_016856355.1 6 988
XM_017000867.1 XP_016856356.1 6 988
XM_017000868.1 XP_016856357.1 6 988
GenBank mRNA AK222754.1 - 6 938
AK295920.1 - 5 774
AK299454.1 - 6 751
AK312416.1 - 6 968
AL050265.1 - 6 954
BC001487.2 - 6 953
BC071657.1 - 6 951 5498250
BC095435.1 - 6 952 30389805
CR533534.1 - 5 866
EF434181.1 - 5 866
EF434182.1 - 5 866
HQ628633.1 - 4 702
HQ628634.1 - 6 775
HQ628635.1 - 5, 6 866
U23731.1 - 5 866

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
transcription from RNA polymerase II promoter
mRNA processing
RNA splicing
negative regulation of gene expression
nuclear fragmentation involved in apoptotic nuclear change
positive regulation of insulin secretion
response to endoplasmic reticulum stress
negative regulation by host of viral transcription
positive regulation of transcription from RNA polymerase II promoter
regulation of cell cycle
3'-UTR-mediated mRNA stabilization
nuclear inner membrane organization
nucleotide binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
mRNA 3'-UTR binding
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline