This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RDH8 SIRNA Assays ›
Gene Symbol:
RDH8
Gene Name:
retinol dehydrogenase 8 (all-trans)
Gene Aliases:
PRRDH, SDR28C2
Chromosome Location:
Chr.19: 10013249 - 10022278 on Build GRCh38
UniGene ID:
Hs.675522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 108766

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RDH8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015725.2 NP_056540.2 6 1476
GenBank mRNA AF229845.1 - 6 1476
AK024022.1 - 6 1241

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
estrogen biosynthetic process
visual perception
retinol metabolic process
response to stimulus
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
retinol dehydrogenase activity
NADP-retinol dehydrogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline