This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other F13B SIRNA Assays ›
Gene Symbol:
F13B
Gene Name:
coagulation factor XIII B chain
Gene Aliases:
FXIIIB
Chromosome Location:
Chr.1: 197038741 - 197067267 on Build GRCh38
UniGene ID:
Hs.435782
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 10905

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F13B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001994.2 NP_001985.2 3 413
XM_011509283.2 XP_011507585.1 3 410
XM_011509284.2 XP_011507586.1 3 410
XM_011509286.2 XP_011507588.1 3 421
GenBank mRNA AK290560.1 - 3 410
BX510295.1 - 3 414
M14057.1 - 3 367

Back To Top

More Information


Gene Ontology Categories:

Function(s)

blood coagulation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-jgmcd:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0