siRNA Details
| Gene Symbol: FCER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207019.2 | NP_001193948.2 | 5 | 368 | ||
| NM_001220500.1 | NP_001207429.1 | 6 | 504 | |||
| NM_002002.4 | NP_001993.2 | 6 | 500 | |||
| XM_005272462.4 | XP_005272519.1 | 6 | 1105 | |||
| GenBank mRNA | AK223207.1 | - | 6 | 469 | ||
| AK310071.1 | - | 6 | 489 | |||
| AK310707.1 | - | 6 | 469 | |||
| BC014108.2 | - | 6 | 465 | 4309266 | ||
| BC062591.1 | - | 6 | 458 | 5216918 | ||
| BC064417.1 | - | 5 | 782 | |||
| M14766.1 | - | 6 | 472 | |||
| M15059.1 | - | 6 | 500 | |||
| X04772.1 | - | 6 | 500 | |||

FCER2
Genomic Map