This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF695 SIRNA Assays ›
Gene Symbol:
ZNF695 ZNF670-ZNF695
Gene Name:
zinc finger protein 695
ZNF670-ZNF695 readthrough (NMD candidate)
Gene Aliases:
SBZF3
-
Chromosome Location:
Chr.1: 246945547 - 247008093 on Build GRCh38
Chromosome Location:
Chr.1: 246945547 - 247078813 on Build GRCh38
UniGene ID:
Hs.669893
Hs.732305
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 109234

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF695
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204221.1 NP_001191150.1 6 860
NR_037892.1 6 864
GenBank mRNA BC023527.2 - 6 772
BC055096.1 - 5 693
BE743952.1 - 5 603
Gene Symbol: ZNF670-ZNF695
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037894.1 7 1025

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0