siRNA Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204456.1 | NP_001191385.1 | 6 | 2884 | ||
| NM_021163.3 | NP_066986.1 | 5 | 3079 | |||
| GenBank mRNA | AK293069.1 | - | 6 | 2884 | ||
| BC136675.1 | - | 5 | 2732 | 9020665 | ||
| BC136676.1 | - | 5 | 2732 | 9020666 | ||

RBAK
Genomic Map