This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRIM34 SIRNA Assays ›
Gene Symbol:
TRIM34 TRIM6-TRIM34
Gene Name:
tripartite motif containing 34
TRIM6-TRIM34 readthrough
Gene Aliases:
IFP1, RNF21
IFP1, RNF21, TRIM34
Chromosome Location:
Chr.11: 5619944 - 5644398 on Build GRCh38
Chromosome Location:
Chr.11: 5596635 - 5644398 on Build GRCh38
UniGene ID:
Hs.729048
Hs.729048
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 109376

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIM34
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003827.1 NP_001003827.1 4, 5 915
NM_021616.5 NP_067629.2 4, 5 919
NM_130390.2 NP_569074.2 3, 4 835
GenBank mRNA AB039902.1 - 4, 5 923
AB039904.1 - 3, 4 817
AF220143.1 - 4, 5 902
AF220144.1 - 5, 6 835
AK027876.1 - 4, 5 891
AL583914.1 - 3, 4 524
Gene Symbol: TRIM6-TRIM34
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003819.3 NP_001003819.1 10, 11 2083
GenBank mRNA AB039903.1 - 10, 11 1991
BC136871.1 - 10, 11 2002 9020862
BC140722.1 - 10, 11 2002 9021590

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of sequence-specific DNA binding transcription factor activity
defense response to virus
interferon-gamma-mediated signaling pathway
protein trimerization
molecular_function
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline