This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FREM1 SIRNA Assays ›
Gene Symbol:
FREM1
Gene Name:
FRAS1 related extracellular matrix 1
Gene Aliases:
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location:
Chr.9: 14734666 - 14911653 on Build GRCh38
UniGene ID:
Hs.50850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 110068

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FREM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177704.1 NP_001171175.1 3 676
NM_144966.5 NP_659403.4 27 5795
XM_005251382.3 XP_005251439.1 26 6043
XM_005251384.4 XP_005251441.1 5 1089
XM_006716729.3 XP_006716792.1 6 1334
XM_011517758.2 XP_011516060.1 3 725
XM_017014316.1 XP_016869805.1 26 6070
XM_017014317.1 XP_016869806.1 27 5613
XM_017014318.1 XP_016869807.1 27 6368
XM_017014319.1 XP_016869808.1 28 6272
XM_017014320.1 XP_016869809.1 27 6199
XM_017014321.1 XP_016869810.1 26 5862
XM_017014322.1 XP_016869811.1 26 5656
XM_017014323.1 XP_016869812.1 26 5329
XM_017014324.1 XP_016869813.1 26 6070
XM_017014325.1 XP_016869814.1 26 6070
XM_017014326.1 XP_016869815.1 25 4886
XM_017014327.1 XP_016869816.1 22 4255
XM_017014328.1 XP_016869817.1 26 6070
XM_017014329.1 XP_016869818.1 26 6070
GenBank mRNA AB160987.1 - 25 4979
AK058190.1 - 2 219
BC031064.1 - 3 676

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell-matrix adhesion
craniofacial suture morphogenesis
carbohydrate binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlc25:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline