This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GOLGA1 SIRNA Assays ›
Gene Symbol:
GOLGA1
Gene Name:
golgin A1
Gene Aliases:
golgin-97
Chromosome Location:
Chr.9: 124878294 - 124941114 on Build GRCh38
UniGene ID:
Hs.133469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 11027

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GOLGA1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002077.3 NP_002068.1 23 4602
XM_005251929.3 XP_005251986.1 23 4534
XM_006717062.3 XP_006717125.1 23 4598
XM_006717063.3 XP_006717126.1 23 4569
GenBank mRNA AB208858.1 - 18 3953
BC032853.1 - 23 4572 4828793
U51587.1 - 23 4592

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to Golgi
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline