This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STK17B SIRNA Assays ›
Gene Symbol:
STK17B
Gene Name:
serine/threonine kinase 17b
Gene Aliases:
DRAK2
Chromosome Location:
Chr.2: 196133583 - 196176503 on Build GRCh38
UniGene ID:
Hs.88297
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 110839

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STK17B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004226.3 NP_004217.1 3 626
XM_011512168.2 XP_011510470.1 3 591
XM_011512169.1 XP_011510471.1 3 624
XM_011512170.1 XP_011510472.1 4 607
XM_011512171.2 XP_011510473.1 1 43
GenBank mRNA AB011421.1 - 3 563
AK312865.1 - 3 624
BC016040.1 - 3 565 4575012
CR542225.1 - 2 302

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
intracellular signal transduction
protein autophosphorylation
positive regulation of fibroblast apoptotic process
protein kinase activity
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-rdx9p:80/100.66.79.90:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline