This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GSTA4 SIRNA Assays ›
Gene Symbol:
GSTA4
Gene Name:
glutathione S-transferase alpha 4
Gene Aliases:
GSTA4-4, GTA4
Chromosome Location:
Chr.6: 52977948 - 52995380 on Build GRCh38
UniGene ID:
Hs.485557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 110958

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSTA4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001512.3 NP_001503.1 7 1049
XM_005249035.3 XP_005249092.1 7 1142
XM_011514534.2 XP_011512836.1 6 992
XM_011514535.2 XP_011512837.1 6 1088
GenBank mRNA AF020918.1 - 7 969
AF025887.1 - 7 972
AK054804.1 - 5 1406
BC015523.2 - 6 919 3915425
BC063439.1 - 5 886 5248427
CR749474.1 - 3 4348

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
xenobiotic metabolic process
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0