siRNA Details
| Gene Symbol: ST6GALNAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320273.1 | NP_001307202.1 | 4 | 1283 | ||
| NM_001320274.1 | NP_001307203.1 | 3 | 1175 | |||
| NM_030965.2 | NP_112227.1 | 5 | 1693 | |||
| GenBank mRNA | AK056241.1 | - | 5 | 1660 | ||
| AK095758.1 | - | 6 | 1579 | |||
| AK300902.1 | - | 4 | 1208 | |||
| BC001201.1 | - | 5 | 1600 | 3356535 | ||

ST6GALNAC5
Genomic Map