This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NDUFA11 SIRNA Assays ›
Gene Symbol:
NDUFA11
Gene Name:
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases:
B14.7, CI-B14.7
Chromosome Location:
Chr.19: 5891276 - 5904013 on Build GRCh38
UniGene ID:
Hs.406062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 112420

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NDUFA11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193375.1 NP_001180304.1 2 467
NM_175614.4 NP_783313.1 2 467
NR_034166.2 3 587
GenBank mRNA AJ539081.1 - 2 162
AK127692.1 - 2 240
AL546175.3 - 3 328
AY726561.1 - 2 223
BC069045.1 - 2 222 6503302
BQ054257.1 - 2 223
BQ671956.1 - 2 241
BQ955016.1 - 2 213

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline