This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRPM2 SIRNA Assays ›
Gene Symbol:
TRPM2
Gene Name:
transient receptor potential cation channel subfamily M member 2
Gene Aliases:
EREG1, KNP3, LTRPC2, LTrpC-2, NUDT9H, NUDT9L1, TRPC7
Chromosome Location:
Chr.21: 44350112 - 44443081 on Build GRCh38
UniGene ID:
Hs.369759
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 113012

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRPM2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320350.1 NP_001307279.1 3 450
NM_001320351.1 NP_001307280.1 3 450
NM_003307.3 NP_003298.1 3 450
NR_038257.1 3 450
XM_005261171.3 XP_005261228.1 4 611
XM_011529736.2 XP_011528038.1 3 854
XM_017028456.1 XP_016883945.1 4 614
XM_017028457.1 XP_016883946.1 3 855
GenBank mRNA AB001535.1 - 3 795
AB166745.1 - 4 563
AJ417076.1 - 3 363
AJ878416.1 - 3 413
AY603182.1 - 3 350
BC112342.1 - 3 363 40069893

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
response to oxidative stress
positive regulation of insulin secretion
sodium ion transmembrane transport
estrous cycle
calcium ion transmembrane transport
calcium ion import across plasma membrane
positive regulation of oxidative stress-induced neuron death
calcium channel activity
sodium channel activity
ADP-ribose diphosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bckb2:80/100.66.73.158:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline