This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSD17B12 SIRNA Assays ›
Gene Symbol:
HSD17B12
Gene Name:
hydroxysteroid 17-beta dehydrogenase 12
Gene Aliases:
KAR, SDR12C1
Chromosome Location:
Chr.11: 43680593 - 43856619 on Build GRCh38
UniGene ID:
Hs.132513
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 113512

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HSD17B12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016142.2 NP_057226.1 5 642
XM_011520156.1 XP_011518458.1 5 636
XM_017017881.1 XP_016873370.1 5 322
GenBank mRNA AF078850.1 - 5 480
AK027882.1 - 535
AK074952.1 - 5 494
AK075011.1 - 5 527
AK075216.1 - 5 503
AK222881.1 - 5 464
AK292625.1 - 5 527
AK301663.1 - 6 433
BC012043.1 - 5 465 3622879

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
long-chain fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
protein binding
collagen binding
heparin binding
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
3-oxo-arachidoyl-CoA reductase activity
3-oxo-behenoyl-CoA reductase activity
3-oxo-lignoceroyl-CoA reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline