siRNA Details
| Gene Symbol: POU4F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006237.3 | NP_006228.3 | 2 | 484 | ||
| GenBank mRNA | L20433.1 | - | 2 | 484 | ||
| X64624.1 | - | 1 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU4F1| Gene Symbol: POU4F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006237.3 | NP_006228.3 | 2 | 484 | ||
| GenBank mRNA | L20433.1 | - | 2 | 484 | ||
| X64624.1 | - | 1 | 275 | |||