This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TFEC SIRNA Assays ›
Gene Symbol:
TFEC
Gene Name:
transcription factor EC
Gene Aliases:
TCFEC, TFE-C, TFEC-L, TFECL, bHLHe34, hTFEC-L
Chromosome Location:
Chr.7: 115935148 - 116159891 on Build GRCh38
UniGene ID:
Hs.125962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 114737

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TFEC
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018058.2 NP_001018068.1 6 724
NM_001244583.1 NP_001231512.1 5 420
NM_012252.3 NP_036384.1 7 811
XM_011515963.1 XP_011514265.1 9 998
XM_011515964.2 XP_011514266.1 9 1013
XM_011515965.2 XP_011514267.1 9 953
XM_011515966.2 XP_011514268.1 9 1062
XM_011515969.1 XP_011514271.1 8 941
XM_011515970.1 XP_011514272.1 8 911
XM_011515972.2 XP_011514274.1 6 527
XM_017011874.1 XP_016867363.1 8 1114
XM_017011875.1 XP_016867364.1 8 949
XM_017011876.1 XP_016867365.1 5 386
XM_017011877.1 XP_016867366.1 7 775
XM_017011878.1 XP_016867367.1 5 313
GenBank mRNA AK301479.1 - 9 1003
AK313546.1 - 7 763
BC029891.1 - 6 655 5179088
CR933605.1 - 6 724
D43945.1 - 7 740

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to heat
negative regulation of nucleic acid-templated transcription
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5wmqj:80/100.66.75.105:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline