This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PGLYRP4 SIRNA Assays ›
Gene Symbol:
PGLYRP4
Gene Name:
peptidoglycan recognition protein 4
Gene Aliases:
PGLYRPIbeta, PGRP-Ibeta, PGRPIB, SBBI67
Chromosome Location:
Chr.1: 153327410 - 153348844 on Build GRCh38
UniGene ID:
Hs.58356
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 114866

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PGLYRP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020393.3 NP_065126.2 9 2062
XM_011509789.2 XP_011508091.1 9 2074
XM_011509791.2 XP_011508093.1 9 2053
XM_011509792.1 XP_011508094.1 9 1754
XM_011509793.2 XP_011508095.1 6 1750
GenBank mRNA AF086392.1 - 1 384
AF242518.1 - 9 1772

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

pattern recognition receptor signaling pathway
peptidoglycan catabolic process
detection of bacterium
innate immune response
defense response to Gram-positive bacterium
zinc ion binding
N-acetylmuramoyl-L-alanine amidase activity
peptidoglycan receptor activity
peptidoglycan binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0