siRNA Details
| Gene Symbol: RING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002931.3 | NP_002922.2 | 7 | 1520 | ||
| GenBank mRNA | AK289355.1 | - | 7 | 1529 | ||
| AK301378.1 | - | 7 | 1463 | |||
| BC002922.2 | - | 7 | 1515 | 3954313 | ||
| BC051866.2 | - | 7 | 1513 | 6142438 | ||
| Z14000.1 | - | 5 | 1314 | |||

RING1
Genomic Map