This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF84 SIRNA Assays ›
Gene Symbol:
ZNF84
Gene Name:
zinc finger protein 84
Gene Aliases:
HPF2
Chromosome Location:
Chr.12: 133037286 - 133063304 on Build GRCh38
UniGene ID:
Hs.445019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115404

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF84
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127372.2 NP_001120844.1 5 980
NM_001289971.1 NP_001276900.1 5 971
NM_001289972.1 NP_001276901.1 5 1099
NM_003428.5 NP_003419.3 5 964
XM_005266185.1 XP_005266242.1 5 1134
XM_005266186.1 XP_005266243.1 6 870
XM_011534832.1 XP_011533134.1 5 914
GenBank mRNA AK314232.1 - 5 742
BC036656.1 - 5 1086 4828620
BC048350.1 - 4 548 5555056
BC062552.1 - 5 740 6536224
M27878.1 - 5 872
X60156.2 - 4 710

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-sqf5z:80/100.66.77.75:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0