This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF84 SIRNA Assays ›
Gene Symbol:
ZNF84
Gene Name:
zinc finger protein 84
Gene Aliases:
HPF2
Chromosome Location:
Chr.12: 133037286 - 133063304 on Build GRCh38
UniGene ID:
Hs.445019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115406

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF84
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127372.2 NP_001120844.1 5 3234
NM_001289971.1 NP_001276900.1 5 3225
NM_001289972.1 NP_001276901.1 5 3353
NM_003428.5 NP_003419.3 5 3218
XM_005266185.1 XP_005266242.1 5 3388
XM_005266186.1 XP_005266243.1 6 3124
XM_011534832.1 XP_011533134.1 5 3168
GenBank mRNA BC036656.1 - 5 3340 4828620
BC048350.1 - 4 2802 5555056
BC062552.1 - 5 2994 6536224
M27878.1 - 5 3125

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline