This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF135 SIRNA Assays ›
Gene Symbol:
ZNF135
Gene Name:
zinc finger protein 135
Gene Aliases:
ZNF61, ZNF78L1, pHZ-17, pT3
Chromosome Location:
Chr.19: 58059227 - 58069755 on Build GRCh38
UniGene ID:
Hs.85863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115412

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF135
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164529.1 NP_001158001.1 4 365
NM_001164530.1 NP_001158002.1 4 724
NM_001289401.1 NP_001276330.1 5 791
NM_001289402.1 NP_001276331.1 5 1154
NM_003436.3 NP_003427.3 5 822
NM_007134.1 NP_009065.1 4 760
XM_005259211.1 XP_005259268.1 4 724
XM_006723362.3 XP_006723425.1 5 830
XM_006723363.3 XP_006723426.1 5 1194
XM_017027240.1 XP_016882729.1 4 1147
XM_017027241.1 XP_016882730.1 5 1149
XM_017027242.1 XP_016882731.1 4 589
GenBank mRNA AK094202.1 - 5 1113
AK098011.1 - 4 724
AK295110.1 - 5 792
AK297226.1 - 4 555
AL833637.1 - 4 323
BC046434.1 - 5 786 4824328
BX647775.1 - 4 728
U09413.1 - 1 193

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cytoskeleton organization
regulation of cell morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline