This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF136 SIRNA Assays ›
Gene Symbol:
ZNF136
Gene Name:
zinc finger protein 136
Gene Aliases:
pHZ-20
Chromosome Location:
Chr.19: 12162672 - 12189880 on Build GRCh38
UniGene ID:
Hs.479874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115416

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF136
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003437.3 NP_003428.1 4 392
XM_011528269.2 XP_011526571.1 5 887
XM_017027243.1 XP_016882732.1 3 650
GenBank mRNA AK123008.1 - 3 1409
AK291242.1 - 4 387
AK299601.1 - 3 215
BC006421.1 - 4 345 4040430
DB200139.1 - 4 392
U09367.1 - 4 316

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
DNA binding
transcription corepressor activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline