This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNHIT3 SIRNA Assays ›
Gene Symbol:
ZNHIT3
Gene Name:
zinc finger HIT-type containing 3
Gene Aliases:
TRIP3
Chromosome Location:
Chr.17: 36486627 - 36499310 on Build GRCh38
UniGene ID:
Hs.2210
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 115563

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNHIT3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281432.1 NP_001268361.1 3 210
NM_001281433.1 NP_001268362.1 3 210
NM_004773.3 NP_004764.1 3 210
NR_104009.1 3 205
NR_104010.1 2 178
NR_104011.1 2 178
GenBank mRNA AB209193.1 - 1 3674
AF400652.1 - 3 202
AK290858.1 - 3 161
AK308583.1 - 2 106
AK310022.1 - 3 145
AV763423.1 - 177
BC017931.1 - 3 175 4669658
BC105608.1 - 2 150
BG032116.1 - 3 145
BI767446.1 - 3 143
BM457955.1 - 140
CB996852.1 - 3 235
HY045632.1 - 3 211
L40410.1 - 3 128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein binding
metal ion binding
thyroid hormone receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline