This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116201

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBM23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077351.1 NP_001070819.1 14 1914
NM_001077352.1 NP_001070820.1 12 1812
NM_001308044.1 NP_001294973.1 12 1753
NM_018107.4 NP_060577.3 13 1866
XM_011536890.2 XP_011535192.1 15 1997
XM_011536892.2 XP_011535194.1 14 1948
XM_011536893.2 XP_011535195.1 13 2029
XM_011536894.2 XP_011535196.1 14 1943
XM_011536895.2 XP_011535197.1 13 1919
XM_011536897.2 XP_011535199.1 13 2281
XM_011536900.1 XP_011535202.1 13 1857
XM_011536902.1 XP_011535204.1 13 1763
XM_011536903.1 XP_011535205.1 11 1639
XM_011536904.1 XP_011535206.1 12 1696
XM_011536905.1 XP_011535207.1 11 1694
XM_011536906.1 XP_011535208.1 10 1585
XM_017021398.1 XP_016876887.1 12 2127
XM_017021399.1 XP_016876888.1 11 2074
XM_017021400.1 XP_016876889.1 11 1992
XM_017021401.1 XP_016876890.1 12 2087
XM_017021402.1 XP_016876891.1 12 1998
XM_017021403.1 XP_016876892.1 11 2016
XM_017021404.1 XP_016876893.1 11 1966
XM_017021405.1 XP_016876894.1 11 2033
XM_017021406.1 XP_016876895.1 14 2329
XM_017021407.1 XP_016876896.1 14 1838
XM_017021408.1 XP_016876897.1 10 1950
XM_017021409.1 XP_016876898.1 13 2275
XM_017021410.1 XP_016876899.1 12 1796
XM_017021411.1 XP_016876900.1 12 2227
XM_017021414.1 XP_016876903.1 11 1939
XM_017021415.1 XP_016876904.1 10 1920
XM_017021416.1 XP_016876905.1 9 1815
XM_017021417.1 XP_016876906.1 10 1872
XM_017021418.1 XP_016876907.1 9 1870
XM_017021419.1 XP_016876908.1 8 1761
XM_017021420.1 XP_016876909.1 11 2114
GenBank mRNA AF087905.1 - 13 1825
AF275678.1 - 14 1874
AK001344.1 - 12 1767
AK225792.1 - 14 1881
AK308055.1 - 13 1831
AL834198.1 - 14 3695
BC002566.2 - 13 1849 3138909
BC024208.2 - 13 1805 2960467
BC106012.1 - 11 1593
BX161440.1 - 14 1847

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
nucleotide binding
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-bwr29:80/100.66.78.44:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0