siRNA Details
| Gene Symbol: ZNF530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321981.1 | NP_001308910.1 | 4 | 694 | ||
| NM_020880.4 | NP_065931.3 | 3 | 642 | |||
| NR_135923.1 | 3 | 642 | ||||
| XM_006723194.1 | XP_006723257.1 | 3 | 641 | |||
| XM_011526919.2 | XP_011525221.1 | 3 | 637 | |||
| XM_011526921.2 | XP_011525223.1 | 4 | 699 | |||
| GenBank mRNA | AB040941.1 | - | 1 | 1293 | ||
| AK096831.1 | - | 3 | 640 | |||
| AL832740.1 | - | 3 | 642 | |||
| BC060865.1 | - | 3 | 642 | 30343224 | ||

ZNF530
Genomic Map