This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF670 SIRNA Assays ›
Gene Symbol:
ZNF670
Gene Name:
zinc finger protein 670
Gene Aliases:
-
Chromosome Location:
Chr.1: 247034638 - 247078813 on Build GRCh38
UniGene ID:
Hs.732305
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 116495

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF670
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204220.1 NP_001191149.1 4 1633
NM_033213.4 NP_149990.1 4 1636
GenBank mRNA AK027325.1 - 1 585
AK074774.1 - 4 1584
BC005360.1 - 4 1583 3684736

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline