This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRIM43 SIRNA Assays ›
Gene Symbol:
TRIM43 TRIM43B
Gene Name:
tripartite motif containing 43
tripartite motif containing 43B
Gene Aliases:
TRIM43A
-
Chromosome Location:
Chr.2: 95592018 - 95599723 on Build GRCh38
Chromosome Location:
Chr.2: 95476967 - 95484731 on Build GRCh38
UniGene ID:
Hs.232026
Hs.723811
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 116554

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIM43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138800.1 NP_620155.1 2 316
GenBank mRNA BC015353.1 - 2 316 3445177
Gene Symbol: TRIM43B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164464.1 NP_001157936.1 2 310
XM_011511669.1 XP_011509971.1 2 254

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Process(es)

protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline