siRNA Details
| Gene Symbol: RNF149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173647.3 | NP_775918.2 | 1, 2 | 592 | ||
| XM_005263920.2 | XP_005263977.1 | 1, 2 | 592 | |||
| XM_005263921.3 | XP_005263978.1 | 1, 2 | 592 | |||
| GenBank mRNA | AK074985.1 | - | 1, 2 | 566 | ||
| AK075141.1 | - | 1, 2 | 566 | |||
| AK129799.1 | - | 1, 2 | 577 | |||
| AY450390.1 | - | 1, 2 | 566 | |||
| BC019355.1 | - | 1, 2 | 543 | 3956746 | ||
| BC032328.1 | - | 1, 2 | 554 | 5106286 | ||
| BC045743.1 | - | 1, 2 | 566 | 5260812 | ||
| DA383672.1 | - | 1, 2 | 592 | |||

RNF149
Genomic Map