siRNA Details
| Gene Symbol: PFDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002623.3 | NP_002614.2 | 4 | 1131 | ||
| GenBank mRNA | U41816.1 | - | 4 | 991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PFDN4| Gene Symbol: PFDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002623.3 | NP_002614.2 | 4 | 1131 | ||
| GenBank mRNA | U41816.1 | - | 4 | 991 | ||