This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SNUPN SIRNA Assays ›
Gene Symbol:
SNUPN
Gene Name:
snurportin 1
Gene Aliases:
KPNBL, RNUT1, Snurportin1
Chromosome Location:
Chr.15: 75598083 - 75626378 on Build GRCh38
UniGene ID:
Hs.21577
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 117371

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNUPN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042581.1 NP_001036046.1 6 857
NM_001042588.1 NP_001036053.1 6 662
NM_005701.3 NP_005692.1 6 962
GenBank mRNA AF039029.1 - 6 607
AK054641.1 - 7 1095
AK289475.1 - 6 658
BC004203.2 - 6 593 3605328
CR456811.1 - 5 517

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal snRNP assembly
protein import into nucleus
nuclear import
snRNA import into nucleus
RNA cap binding
protein transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-2d48v:80/100.66.79.208:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0