This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CEP41 SIRNA Assays ›
Gene Symbol:
CEP41
Gene Name:
centrosomal protein 41
Gene Aliases:
JBTS15, TSGA14
Chromosome Location:
Chr.7: 130393771 - 130441210 on Build GRCh38
UniGene ID:
Hs.368315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 117993

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CEP41
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257158.1 NP_001244087.1 2 324
NM_001257159.1 NP_001244088.1 2 324
NM_001257160.1 NP_001244089.1 2 324
NM_018718.2 NP_061188.1 2 324
NR_046443.1 2 324
XM_011516709.2 XP_011515011.1 2 171
GenBank mRNA AF429308.1 - 2 170
AF429309.1 - 2 170
AJ278890.1 - 2 127
AK026098.1 - 2 124
AK298618.1 - 2 140
AK311382.1 - 2 141
AK314676.1 - 2 103
BC018789.1 - 2 106
BC056162.1 - 2 97 6142659
CB155730.1 - 2 351
DB069500.1 - 2 326

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
protein transport
single organismal cell-cell adhesion
protein polyglutamylation
cilium assembly
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline