This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PCK2 SIRNA Assays ›
Gene Symbol:
PCK2
Gene Name:
phosphoenolpyruvate carboxykinase 2, mitochondrial
Gene Aliases:
PEPCK, PEPCK-M, PEPCK2
Chromosome Location:
Chr.14: 24094131 - 24104132 on Build GRCh38
UniGene ID:
Hs.75812
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 118728

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PCK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018073.2 NP_001018083.2 6, 7 1284
NM_001291556.1 NP_001278485.1 6, 7 1118
NM_001308054.1 NP_001294983.1 6, 7 1545
NM_004563.3 NP_004554.3 6, 7 1284
XM_006720158.2 XP_006720221.1 6, 7 1277
GenBank mRNA AK129934.1 - 6, 7 1102
AK301400.1 - 6, 7 1128
AK316206.1 - 6, 7 938
BC001454.2 - 6, 7 1074 3138368
BX248272.1 - 6, 7 1058
CR456913.1 - 6, 7 1009
KU178243.1 - 6, 7 1009
KU178244.1 - 6, 7 1009
X92720.1 - 6, 7 1075

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
phosphoenolpyruvate carboxykinase activity
phosphoenolpyruvate carboxykinase (GTP) activity
protein binding
GTP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0