This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PEX1 SIRNA Assays ›
Gene Symbol:
PEX1
Gene Name:
peroxisomal biogenesis factor 1
Gene Aliases:
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
Chromosome Location:
Chr.7: 92487023 - 92528531 on Build GRCh38
UniGene ID:
Hs.164682
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 119067

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PEX1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000466.2 NP_000457.1 2 240
NM_001282677.1 NP_001269606.1 2 240
NM_001282678.1 NP_001269607.1 2 240
GenBank mRNA AB008112.1 - 2 211
AB052090.1 - 2 144
AB052091.1 - 2 144
AB052092.1 - 2 144
AB052093.1 - 2 144
AB052094.1 - 2 144
AF026086.1 - 2 204
AF030356.1 - 2 148
AK292955.1 - 2 145
AK293757.1 - 2 148
AK295686.1 - 2 150
BC035575.1 - 2 182 5498274
DA313282.1 - 2 241
DC370317.1 - 2 216

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to peroxisome
peroxisome organization
protein import into peroxisome matrix
microtubule-based peroxisome localization
protein binding
ATP binding
protein C-terminus binding
protein complex binding
ATPase activity, coupled

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-mmd6l:80/100.66.75.105:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0