siRNA Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024758.4 | NP_079034.3 | 2 | 520 | ||
| GenBank mRNA | AK130034.1 | - | 2 | 316 | ||
| AK225366.1 | - | 2 | 433 | |||
| AY057097.1 | - | 2 | 420 | |||
| BC005090.1 | - | 2 | 520 | 3837251 | ||
| BM788254.1 | - | 2 | 451 | |||
| DN996958.1 | - | 2 | 510 | |||

AGMAT
Genomic Map