siRNA Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024758.4 | NP_079034.3 | 7 | 1379 | ||
| GenBank mRNA | AK130034.1 | - | 7 | 1175 | ||
| AY057097.1 | - | 7 | 1279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGMAT| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024758.4 | NP_079034.3 | 7 | 1379 | ||
| GenBank mRNA | AK130034.1 | - | 7 | 1175 | ||
| AY057097.1 | - | 7 | 1279 | |||