This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSG11 SIRNA Assays ›
Gene Symbol:
PSG11 PSG10P PSG5
Gene Name:
pregnancy specific beta-1-glycoprotein 11
pregnancy specific beta-1-glycoprotein 10, pseudogene
pregnancy specific beta-1-glycoprotein 5
Gene Aliases:
PSBG-11, PSBG-13, PSG13, PSG14
PSG10, PSG12
FL-NCA-3, PSG
Chromosome Location:
Chr.19: 43007657 - 43026479 on Build GRCh38
Chromosome Location:
Chr.19: 42836997 - 42855718 on Build GRCh38
Chromosome Location:
Chr.19: 43167743 - 43186536 on Build GRCh38
UniGene ID:
Hs.741150
Hs.655399
Hs.654415
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 12030

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PSG11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113410.1 NP_001106881.1 4, 5 797
NM_002785.2 NP_002776.3 5, 6 1163
GenBank mRNA BE767872.1 - 2, 3 266
BG571293.1 - 3, 4 505
M69245.1 - 5, 6 1125
U25988.1 - 4, 5 793
Gene Symbol: PSG10P
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026824.1 6, 7 1364
Gene Symbol: PSG5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130014.1 NP_001123486.1 5, 6 1187
GenBank mRNA M25384.1 - 6, 7 1155
M73713.1 - 5, 6 1141

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

female pregnancy
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline