This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RAB20 SIRNA Assays ›
Gene Symbol:
RAB20
Gene Name:
RAB20, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.13: 110523066 - 110561737 on Build GRCh38
UniGene ID:
Hs.743563
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 120588

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RAB20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017817.2 NP_060287.1 2 462
GenBank mRNA AJ272065.1 - 2 244
AK000436.1 - 2 449
AK000446.1 - 2 449
BC026025.1 - 2 458 4636491
BU528103.1 - 462
HY010547.1 - 2 417

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
protein transport
phagosome acidification
phagosome-lysosome fusion
regulation of autophagosome assembly
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-9n26k:80/100.66.76.30:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline