This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HDAC10 SIRNA Assays ›
Gene Symbol:
HDAC10
Gene Name:
histone deacetylase 10
Gene Aliases:
HD10
Chromosome Location:
Chr.22: 50245183 - 50251405 on Build GRCh38
UniGene ID:
Hs.26593
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 120681

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HDAC10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159286.1 NP_001152758.1 5 855
NM_032019.5 NP_114408.3 5 855
GenBank mRNA AF393962.1 - 5 482
AF407272.1 - 5 482
AF407273.1 - 5 482
AF426160.1 - 5 835
AY450395.1 - 5 482
BC032049.1 - 5 668
BC094734.1 - 4 614
BC125082.1 - 4 432
BC125083.1 - 5 529 40117772
CR456465.1 - 5 538

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
oligodendrocyte development
chromatin modification
histone deacetylation
negative regulation of transcription, DNA-templated
histone H3 deacetylation
histone deacetylase activity
protein binding
enzyme binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline