siRNA Details
| Gene Symbol: OSBPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145798.2 | NP_665741.1 | 4 | 433 | ||
| XM_017024122.1 | XP_016879611.1 | 4 | 455 | |||
| XM_017024123.1 | XP_016879612.1 | 4 | 573 | |||
| GenBank mRNA | AF323729.1 | - | 4 | 422 | ||
| AF392446.1 | - | 4 | 486 | |||
| AK289523.1 | - | 4 | 455 | |||
| BC028010.1 | - | 4 | 432 | |||
| BC065482.1 | - | 4 | 405 | 6081784 | ||

OSBPL7
Genomic Map