This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TONSL SIRNA Assays ›
Gene Symbol:
TONSL
Gene Name:
tonsoku-like, DNA repair protein
Gene Aliases:
IKBR, NFKBIL2
Chromosome Location:
Chr.8: 144428780 - 144444488 on Build GRCh38
UniGene ID:
Hs.675285
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 121665

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TONSL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013432.4 NP_038460.4 6 608
XM_011517050.2 XP_011515352.1 6 667
GenBank mRNA AB209126.1 - 1 459
BC008782.2 - 5 580 3628374

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
replication fork processing
cytoplasmic sequestering of transcription factor
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
protein binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9w28n:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline