siRNA Details
| Gene Symbol: ANGPTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178127.4 | NP_835228.2 | 9 | 2235 | ||
| XM_011542735.2 | XP_011541037.1 | 7 | 2034 | |||
| XM_017017466.1 | XP_016872955.1 | 5 | 1809 | |||
| GenBank mRNA | AY169281.1 | - | 8 | 1693 | ||
| AY358827.1 | - | 8 | 1683 | |||
| BC049170.1 | - | 9 | 2238 | 5285447 | ||
| BC053957.1 | - | 9 | 2238 | |||

ANGPTL5
Genomic Map