This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ECD SIRNA Assays ›
Gene Symbol:
ECD
Gene Name:
ecdysoneless cell cycle regulator
Gene Aliases:
GCR2, HSGT1, SGT1
Chromosome Location:
Chr.10: 73134524 - 73168095 on Build GRCh38
UniGene ID:
Hs.631822
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 122626

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ECD
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135752.1 NP_001129224.1 7 1140
NM_007265.2 NP_009196.1 7 1140
NR_024203.1 5 729
GenBank mRNA AK022616.1 - 7 1088
AK315711.1 - 7 1113
BC000721.2 - 7 1104 3504956
CR457407.1 - 6 897
D88208.1 - 7 1104

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of glycolytic process
transcription from RNA polymerase II promoter
mRNA processing
cell proliferation
RNA splicing
positive regulation of transcription from RNA polymerase II promoter
regulation of G1/S transition of mitotic cell cycle
transcription coactivator activity
protein binding
histone acetyltransferase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline