This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C11ORF49 SIRNA Assays ›
Gene Symbol:
C11orf49
Gene Name:
chromosome 11 open reading frame 49
Gene Aliases:
-
Chromosome Location:
Chr.11: 46936524 - 47164381 on Build GRCh38
UniGene ID:
Hs.368296
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 123385

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C11orf49
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003676.2 NP_001003676.1 7 826
NM_001003677.2 NP_001003677.1 7 826
NM_001003678.2 NP_001003678.1 7 826
NM_001278222.1 NP_001265151.1 6 723
NM_024113.4 NP_077018.1 7 826
NR_103471.1 8 1071
NR_103472.1 6 695
XM_006718315.1 XP_006718378.1 7 817
XM_011520364.2 XP_011518666.1 6 888
XM_011520365.2 XP_011518667.1 7 817
XM_017018276.1 XP_016873765.1 7 817
XM_017018277.1 XP_016873766.1 6 636
XM_017018278.1 XP_016873767.1 8 1000
XM_017018279.1 XP_016873768.1 8 1063
XM_017018280.1 XP_016873769.1 9 1724
XM_017018281.1 XP_016873770.1 8 1693
XM_017018282.1 XP_016873771.1 8 1000
GenBank mRNA AK025863.1 - 7 735
AK293609.1 - 6 604
AK300813.1 - 6 633
AK315639.1 - 7 774
AK316318.1 - 8 1063
AL136575.1 - 7 745
BC001860.2 - 7 772 3534541
BC009455.1 - 7 747 3535652
BC013988.2 - 7 737 3349493
BC052291.1 - 7 739 5936112
CR457351.1 - 7 709

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline