This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HARBI1 SIRNA Assays ›
Gene Symbol:
HARBI1
Gene Name:
harbinger transposase derived 1
Gene Aliases:
C11orf77
Chromosome Location:
Chr.11: 46603027 - 46617651 on Build GRCh38
UniGene ID:
Hs.714463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 124247

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HARBI1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173811.3 NP_776172.1 2 325
XM_011520025.1 XP_011518327.1 2 325
XM_011520026.2 XP_011518328.1 2 302
XM_011520027.1 XP_011518329.1 2 336
XM_011520028.2 XP_011518330.1 2 311
XM_011520029.2 XP_011518331.1 2 597
XM_017017602.1 XP_016873091.1 2 441
XM_017017603.1 XP_016873092.1 2 356
XM_017017604.1 XP_016873093.1 2 336
XM_017017605.1 XP_016873094.1 2 597
XM_017017606.1 XP_016873095.1 2 352
GenBank mRNA AK057237.1 - 2 325
BC036925.1 - 2 257 5225925

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001113793, XM_006234587, XM_006234588, BC158734.
This siRNA also targets the following transcripts in Mouse: NM_178724, BC094315, AK045343, AK080671, AK084226, and others.

Gene Ontology Categories:

Function(s) Process(es)

nucleic acid phosphodiester bond hydrolysis
nuclease activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0