This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STPG1 SIRNA Assays ›
Gene Symbol:
STPG1
Gene Name:
sperm tail PG-rich repeat containing 1
Gene Aliases:
C1orf201, MAPO2
Chromosome Location:
Chr.1: 24356999 - 24415533 on Build GRCh38
UniGene ID:
Hs.403187
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 124305

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STPG1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199012.1 NP_001185941.1 6, 7 834
NM_001199013.1 NP_001185942.1 6, 7 746
NM_001199014.1 NP_001185943.1 7, 8 890
NM_178122.4 NP_835223.1 5, 6 627
XM_006711025.2 XP_006711088.1 7, 8 852
XM_011542403.2 XP_011540705.1 5, 6 1151
XM_011542404.1 XP_011540706.1 7, 8 965
XM_017002768.1 XP_016858257.1 7, 8 961
XM_017002769.1 XP_016858258.1 8, 9 1109
XM_017002770.1 XP_016858259.1 9, 10 1224
GenBank mRNA AK025957.1 - 4, 5 557
AK299401.1 - 6, 7 600
BC017650.1 - 2, 3 153
BC035061.1 - 7, 8 863 5262741
BC047705.1 - 4, 5 532 5764388
BC063891.1 - 6, 7 778 30390664
BM922375.1 - 6, 7 709

Back To Top

More Information


Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s)

apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4zngx:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline