This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SDHB SIRNA Assays ›
Gene Symbol:
SDHB
Gene Name:
succinate dehydrogenase complex iron sulfur subunit B
Gene Aliases:
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP
Chromosome Location:
Chr.1: 17018722 - 17054170 on Build GRCh38
UniGene ID:
Hs.465924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 12435

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SDHB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003000.2 NP_002991.2 2 312
GenBank mRNA AK312056.1 - 2 174
BC007840.2 - 2 312 4124737
BF796275.1 - 2 160
D10245.1 - 1 107
DQ403007.1 - 2 134
M32246.1 - 1 107
U17248.1 - 2 295

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
succinate metabolic process
aerobic respiration
respiratory electron transport chain
protein binding
succinate dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
ubiquinone binding
2 iron, 2 sulfur cluster binding
3 iron, 4 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ln7zh:80/100.66.79.20:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0